Standard storage cabinets, Protector®
Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel, Singler right hinge, 610×560×900 mm, 59 kg
Catalog # LBCP9900300
Supplier: LABCONCO
Jump to:
Product Details & Documents
Storage cabinets support fume hoods and used for storage and venting of small laboratory instruments, mild acids and other reagents.
- Manual-closing, non-locking doors
- Durable epoxy coated steel construction
Delivery information: Includes four leveling feet and one filter panel (20 cm) to increase cabinet depth from 55 to 76,2 cm. Spillstopper™ work surfaces, flat work surfaces, shelf kits and filter panels can be ordered separately.
Specifications
- Description:Protector Standard Storage Cabinets support Labconco fume hoods Protector®, with filter panel
- Seq:01
- W×D×H:610×560×900 mm
- Doors:Singler right hinge
- Weight:59 kg
Accessories
Product Information
- Catalog #AvailabilityPrice
- Interior epoxy coated shelf and mounting hardware, 762 mm width, 4,5 kgInterior epoxy coated shelf and mounting hardware, 762 mm width, 4,5 kgLBCP9919102Log in for availability1 * 1 items



