Safety spectacles, Contour
Anti-UV 2C-1.2, anti-scratch, anti-fog, Grey, Clear PC
Catalog # 111-0112
Supplier: Bollé Safety
Jump to:
Product Details & Documents
Solidity, protection, design, comfort and lightness make Contour safety glasses a unique model.
- Non-slip TIPGRIP temples
- Non-slip bridge
- Model with ESP (Extra Sensory Perception) lens: An innovative coating to reduce dazzle and the harmful effects of blue light
Weight: 21 g
Certifications: EN 166, (EN 170, EN 172)
Delivery information: Supplied with microfibre pouch.
Specifications
- Type:Anti-UV 2C-1.2, anti-scratch, anti-fog
- Colour:Grey
- Seq:01
- Lenses:Clear PC