Order Entry
Export
Orders LinkContactUsLinkComponent
pH buffers, ready to use

pH buffers, ready to use

Calibration and maintenance set with technical buffer solution for gel electrodes (SORT/TPL/G/TRACE)
3 bottles with each 250 ml: pH 4,01/7,0/10,01 Trace,
2 bottles with each 250 ml KCl solution 3 mol/L, Plastic dosing bottle, 1 SET
Catalog # WTWA108825
Supplier:   WTW
pH buffers, ready to use
Catalog # WTWA108825
Supplier:  WTW
CAS Number:  
pH buffers, ready to use
pH buffers, ready to use

Calibration and maintenance set with technical buffer solution for gel electrodes (SORT/TPL/G/TRACE)
3 bottles with each 250 ml: pH 4,01/7,0/10,01 Trace,
2 bottles with each 250 ml KCl solution 3 mol/L, Plastic dosing bottle, 1 SET

CAS Number:  
Price
93,00 €
Log in for availability
1 * 1 SET
Product Details & Documents

Every bottle has its own lot reference with the expiration date.

  • Shelf life up to 3 years (unopened)
  • Reference temperature at 25 °C
  • Economic consumption
  • Easiest handling by use of the dosing vessel
  • One litre container with a separate compartment for the sample

Accuracy is ±0,03 pH

Certifications: Traceable to NIST.

Specifications

Specifications

  • Pack type:
    Plastic dosing bottle
  • Pk:
    1 SET
  • Description:
    Calibration and maintenance set with technical buffer solution for gel electrodes (SORT/TPL/G/TRACE)<br />3 bottles with each 250 ml: pH 4,01/7,0/10,01 Trace,<br />2 bottles with each 250 ml KCl solution 3 mol/L

Specifications