ContactUsLinkComponent
Areometer, DIN12791, M50 series, measuring range: 1.30-1.35:0.0010g/cm , without thermometer, reference temperature:20°C, length:270 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH808028
Supplier:  Amarell
Areometer, DIN12791, M50 series, measuring range: 1.30-1.35:0.0010g/cm , without thermometer, reference temperature:20°C, length:270 mm, suitable for government verification. 1 * 1 items
Catalog # AMARH808028
Supplier:  Amarell