Overalls, Tritex® pro, SMS
White, XL
113-0655
Jump to:
This overall made from three ply SMS fabric is robust, breathable and anti-static. It offers comfort in all work areas.
- Hood with elastic tie
- Zip covering strip
- Elasticated waist, sleeves and legs
Suitable for production and bottling activities in the food, pharmaceutical or cosmetic industries as well as for production and processing in the chemical and veterinarian industries etc.
Colour: White.
Certifications: CE 0624, Cat. III, Type 5/6
Specifications
- Clothing Size:XL
- Colour:White
- Seq:02
- Cat. No.:113-0655
- Anti-static:+
- Garment application:Laboratory|Food