Orders LinkContactUsLinkComponent
Double spatulas, micro, American pattern
Double spatulas, micro, American pattern
Double ended, American pattern, 160 mm
Catalog # HAMMHSN417-16
Supplier:   HAMMACHER KARL
Double spatulas, micro, American pattern
Catalog # HAMMHSN417-16
Supplier:  HAMMACHER KARL
Double spatulas, micro, American pattern
Double spatulas, micro, American pattern
Double ended, American pattern, 160 mm
Price
10,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These double-ended micro spatulas with American pattern are made of stainless steel.
Specifications
  • Description:
    Double ended, American pattern
  • Length:
    160 mm
  • Seq:
    02