Orders LinkContactUsLinkComponent
Instrument dishes
Lids for instrument trays 2300/5500 ml, PS crystal clear, 360×260×20 mm
  BURK4230-2231
 :   BURKLE
Instrument dishes
  BURK4230-2231
 :  BURKLE
Instrument dishes
Instrument dishes
Lids for instrument trays 2300/5500 ml, PS crystal clear, 360×260×20 mm
18,50 €
1 * 1 items
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
  • Melamine, white
  • Tray without lid
Lid available as accessory.
  • Description:
    Lids for instrument trays 2300/5500 ml, PS crystal clear
  • W×D×H:
    360×260×20 mm