Ethanol ≥96% (v/v), TechniSolv®, pure
Plastic receptacle, Duty paid, 5 L
Ethanol ≥96% (v/v), TechniSolv®, pure
Catalog # 83804.360_T
CAS Number: 64-17-5
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Ethanol 96 % vol pure TechniSolv 1 * 20 L Catalog #83804.440
Product 83804.440 is similar to 83804.360_T but may have a different manufacturer and/or packaging.
Ethanol 96 % vol pure TechniSolv 1 * 10 L Catalog #83804.410
Product 83804.410 is similar to 83804.360_T but may have a different manufacturer and/or packaging.
Product Details & Documents
Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Boiling Pt:78.3 °C (1013 hPa)
- Melting Pt:–117 °C
- Density:0.7895 g/cm³ (20 °C)
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-578-6
- UN:1170
- ADR:3,II
- REACH:01-2119457610-43
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
P20_VWRC83804.360_SPEC_EN.pdf P20_VWRC83804.410_SPEC_EN.pdf P20_VWRC83804.440_SPEC_EN.pdf P20_VWRC83804.550_SPEC_EN.pdf spec_VWRC83804.360
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Specifications
Specification Test Results
- Assay:Min. 95 %
- Assay (V/V):Min. 95 %
- Density (20/20):0.805 - 0.811
Specifications
- Pack type:Plastic receptacle
- Pk:5 L
- Duty status:Duty paid



