Orders LinkContactUsLinkComponent
Agarose, MetaPhor™
Agarose, MetaPhor™
Agarose, MetaPhor™
Catalog # 733-1212
Supplier:   LONZA
Agarose, MetaPhor™
Catalog # 733-1212
Supplier:  LONZA
Agarose, MetaPhor™
Agarose, MetaPhor™
Agarose, MetaPhor™
Price
3 210,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Intermediate melting point agarose ideal for resolution of DNA fragments, PCR and RT-PCR products, differing in size by 2%, in the range of 20 to 800 bp, by submarine gel electrophoresis.
MetaPhor™ agarose gel is ideal for resolving AMPFLPs, STRs, and tri- and tetranucleotide repeats.
Specifications
  • Description:
    Agarose, MetaPhor™
  • Seq:
    03