n-Dodecane
1
Catalog # METN51338012
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
DODECAN Catalog #METT51338012
Product METT51338012 is a replacement for METN51338012 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Synonyms: Dodecane
- CAS Number:112-40-3
- Formula:H₃C(CH₂)₁₀CH₃
- MDL Number:MFCD00008969
- Molecular Weight:170.34 g/mol
- Boiling Pt:216 °C (1013 hPa)
- Melting Pt:–9.6 °C
- Density:0.7487 g/cm³ (20 °C)
- Flash Pt:74 °C
- Storage Temperature:Ambient
- EINECS:203-967-9
Specifications
Specifications
- Pk:1



