Mini bottle-top dispensers, MINISPENSOR
1000 - 2000 µl, ≤±0,7 %, <0,2 %
Catalog # WITG5.371.990
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
This fix volume bottle dispenser is efficient for accurate delivery of microliter quantities.
- For single handed operation
- Suitable for safe storage in a fridge
- Substitution for microliter pipettes
These dispenser are used for hospital laboratories for various tests like glucose, gama GT, creatinine, urea, cholesterol, amylase, lipose, iric acid, bilirubin.
: Supplied with a thread connector GL 28, 1 volume adapter and 1 bottle, both NS 29/32, suction tube and discharge tube.
Specifications
- Volume:1000 - 2000 µl
- Seq:04
- Accuracy:≤±0,7 %
- Imprecision:<0,2 %
Accessories
Product Information
- Catalog #AvailabilityPrice
- Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, round form, 2500 ml, thread GL 45Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, round form, 2500 ml, thread GL 45661-3194Log in for availability1 * 1 items - 24,80 €
Product Information
- Catalog #AvailabilityPrice
- Check valve in borosilicate glass with plastic coatingCheck valve in borosilicate glass with plastic coating613-9192Log in for availability1 * 1 items - 25,60 €
Product Information
- Catalog #AvailabilityPrice
- Calcium chloride tube in borosilicate glass with plastic coatingCalcium chloride tube in borosilicate glass with plastic coating613-9191Log in for availability1 * 1 items - 10,00 €
Product Information
- Catalog #AvailabilityPrice
- Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 1000 ml, thread GL 45Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 1000 ml, thread GL 45661-3192Log in for availability1 * 1 items - 12,70 €
Product Information
- Catalog #AvailabilityPrice
- Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 500 ml, thread GL 32Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 500 ml, thread GL 32215-1714Log in for availability1 * 1 items - 10,00 €
Product Information
- Catalog #AvailabilityPrice
- Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 250 ml, thread GL 32Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 250 ml, thread GL 32215-1712Log in for availability1 * 1 items - 9,70 €
Product Information
- Catalog #AvailabilityPrice
- Thread adapter in PP, A 32 / A 28
Product Information
- Catalog #AvailabilityPrice
- Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 100 ml, thread GL 32Bottle in soda lime brown-glass, LEVASINT®-coated, without screw cap, 100 ml, thread GL 32215-1710Log in for availability1 * 1 items - 7,80 €
Product Information
- Catalog #AvailabilityPrice
- Bottle (Minispensor) in soda lime brown-glass, LEVASINT®-coated, without screw cap, 100 ml, thread GL 28Bottle (Minispensor) in soda lime brown-glass, LEVASINT®-coated, without screw cap, 100 ml, thread GL 28WITG5.372.128Log in for availability1 * 1 items - 5,10 €