Double bossheads
With nickel plated brass screws, Zinc die-cast, powder coated, 20,0 mm
Catalog # 241-7104
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
For stand assembly.
- Quick, easy assembly and disassembly
- Robust, durable
- Wide range of different materials available for many applications
Specifications
- Description:With nickel plated brass screws
- Material:Zinc die-cast, powder coated
- Seq:4
- For rod Ø:20,0 mm