Orders LinkContactUsLinkComponent
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 6% : 0,1
Catalog # 709-0142
Supplier:   GERBER FUNKE
Milk butyrometer
Catalog # 709-0142
Supplier:  GERBER FUNKE
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 6% : 0,1
Price
3,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specifications
  • Description:
    Butyrometer for milk, 0 - 6% : 0,1
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, conical, for all measuring method butyrometers
    Stopper, rubber, conical, for all measuring method butyrometers
    614-2028
    Log in for availability
    1 * 100 items - 48,20 €