Orders LinkContactUsLinkComponent
Lids for Nunc™ microplates
Lid, PS, with cut-off corners, condensation rings and evaporation barrier, for use with 96 MicroWell™ plates, Clear, 7,9 mm, Sterile
  392-0395
   Low Manufacturing Impact
Lids for Nunc™ microplates
  392-0395
   Low Manufacturing Impact
Lids for Nunc™ microplates
Lids for Nunc™ microplates
Lid, PS, with cut-off corners, condensation rings and evaporation barrier, for use with 96 MicroWell™ plates, Clear, 7,9 mm, Sterile
140,00 €
1 * 60 items
  • Sterility:
    Sterile
  • Colour:
    Clear
  • Description:
    Lid, PS, with cut-off corners, condensation rings and evaporation barrier, for use with 96 MicroWell™ plates
  • Height:
    7,9 mm
  • Seq:
    06
  • Cat. No.:
    392-0395
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact