Order Entry
Export
Orders LinkContactUsLinkComponent
Disposable sack masters, SteriWare®

Disposable sack masters, SteriWare®

Disposable sack master, HDPE, 25 mm, 400 mm
Catalog # 231-3504
Supplier:   Sampling Systems
Disposable sack masters, SteriWare®
Catalog # 231-3504
Supplier:  Sampling Systems
Disposable sack masters, SteriWare®
Disposable sack masters, SteriWare®

Disposable sack master, HDPE, 25 mm, 400 mm

Price
421,00 €
Log in for availability
1 * 100 items
Product Details & Documents

These disposable sack masters are hygienic single use devices.

Specially designed to take samples through the side of a sack.

Packaging: Individually bagged.

Specifications
  • Ø:
    25 mm
  • Description:
    Disposable sack master
  • Length:
    400 mm
  • Material:
    HDPE
Accessories

Product Information

  • Catalog #
    Capacity
    L×W
    Availability
    Price
  • Pure Bag, leak proof
    Pure Bag, leak proof
    SAMP8080A-450
    450 ml
    229×114 mm
    Log in for availability
    1 * 1.000 items - 509,00 €