Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Closures
Test/Sample Tube Closures
Stoppers for tubes
Stoppers for tubes
Print
Stoppers for tubes
Grip stopper, 16 mm O.D., Natural
Catalog # 212-0642
Supplier:
Greiner Bio-One
Stoppers for tubes
Catalog # 212-0642
Supplier:
Greiner Bio-One
Expand image
Return to Top
Stoppers for tubes
Grip stopper, 16 mm O.D., Natural
Specifications
Add to cart
Jump to:
Specifications
Price
4,80 €
Log in
for availability
Unit of Measure
1 * 100 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
For tubes:
16 mm O.D.
Colour:
Natural
Description:
Grip stopper
Seq:
07