Orders LinkContactUsLinkComponent
Accessories for pH electrodes
Accessories for pH electrodes
Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
Catalog # WTWA109735
Supplier:   WTW
Accessories for pH electrodes
Catalog # WTWA109735
Supplier:  WTW
Accessories for pH electrodes
Accessories for pH electrodes
Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
Price
53,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Documents
Specifications
  • Description:
    Electrolyte with 2 mol/L KNO₃ and 0,001 mol/L KCl for combined ORP electrode with silver electrode
  • Seq:
    06