0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Closures
Bottle Closures
Screw Caps for Bottles
Screw Caps for Bottles
Screw caps with foam inlay, GL 40, Black, PP
RIXI8-1002-40
:
BERLIN PACKAGING RIXIUS GMBH
Screw Caps for Bottles
RIXI8-1002-40
:
BERLIN PACKAGING RIXIUS GMBH
10+
Screw Caps for Bottles
Screw caps with foam inlay, GL 40, Black, PP
14,00 €
1 * 25 items
-
+
Thread:
GL 40
Colour:
Black
Description:
Screw caps with foam inlay
Material:
PP
Seq:
02