Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REACTION VESSEL 2.0ML 1000µL OPEN END 1 * 1 items
Catalog # DIAS920535
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
Expand image
REACTION VESSEL 2.0ML 1000µL OPEN END 1 * 1 items
Catalog # DIAS920535
Supplier:
DIAGNOSTIC SYSTEMS DIASYS