Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REACTION VESSEL 2.0ML 1000µL END TO END 1 * 1 items
Catalog # DIAS920534
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
Expand image
REACTION VESSEL 2.0ML 1000µL END TO END 1 * 1 items
Catalog # DIAS920534
Supplier:
DIAGNOSTIC SYSTEMS DIASYS