Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Dispenser Tips, Universal

25,0 ml, +, Individual blister
  613-1032
VWR®, Dispenser Tips, Universal
  613-1032
VWR®, Dispenser Tips, Universal
VWR®, Dispenser Tips, Universal

25,0 ml, +, Individual blister

 
 
   
1 * 25 items
 

By using the direct displacement method, these tips work well for infectious and volatile liquids, as the fluid never will get in touch with the instrument which avoids any contamination.

  • Compatible with: VWR® Stepper and Stepper Pro, Ritter Ripette®, and Ripette® pro, Brand HandyStep®, HandyStep® electronic and HandyStep® touch*, Eppendorf® Multipette® 4780 and Multipette® plus, Gilson Repetman®, EasyStep, Minilab 100/101, STEPMATE Handrop (* without activated display/tip recognition)
  • Sustainable design based on virgin resins
  • Nine different tip sizes from 0,1 to 50 ml
  • Standard (non sterile, but PCR-clean = ATP-, RNase-, DNase- and endotoxin-free) with sterile tips available
  • Sterile tips guaranteed free from DNA, DNase, RNase, ATP and pyrogens

NOTE: 25 and 50 ml tips require an adapter, which is included in the delivery.

 
  • Packed:
    Individual blister
  • Sterile:
    +
  • Volume:
    25,0 ml
  • Seq:
    20