Connection system GL 45, DURAN®
Screw cap, GL 45, PP, with three ports GL 14
Catalog # 554-3001
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Connection system with PP screw cap GL 45 with two or three PTFE ports. Flexible modular system. Typical applications: safe transfer of liquid media within a closed and sterile system (evaporation is reduced).
- Four tubing sizes (1,6, 3,0, 3,2 or 6,0 mm) can be used by selecting the correct insert
- Sterile pressure equalisation is possible through use of membrane filter
- Unused ports can be provided with a blind cap
Documents
SCOT1129750_TDS_EN
Datasheet
Specifications
- Description:Screw cap, GL 45, PP, with three ports GL 14
Accessories
Product Information
- Catalog #AvailabilityPrice
- Screw cap with bore, for tube connection, GL 14, blueScrew cap with bore, for tube connection, GL 14, blue554-3004Log in for availability1 * 2 items - 9,50 €
Product Information
- Catalog #AvailabilityPrice
- Insert for screw cap GL 14, 1,6 mm inner diameterInsert for screw cap GL 14, 1,6 mm inner diameter554-3005Log in for availability1 * 1 items - 15,80 €
Product Information
- Catalog #AvailabilityPrice
- Screw cap, closed , with seal, GL 14, red
Product Information
- Catalog #AvailabilityPrice
- Insert for screw cap GL 14, 3,0 mm inner diameterInsert for screw cap GL 14, 3,0 mm inner diameter554-3006Log in for availability1 * 1 items - 15,80 €
Product Information
- Catalog #AvailabilityPrice
- Insert for screw cap GL 14, 3,2 mm inner diameterInsert for screw cap GL 14, 3,2 mm inner diameter554-3007Log in for availability1 * 1 items - 15,80 €
Product Information
- Catalog #AvailabilityPrice
- Pressure equalization set for 2 and 3 port screw cap, incl. 0,2 μm membrane filterPressure equalization set for 2 and 3 port screw cap, incl. 0,2 μm membrane filter554-3009Log in for availability1 * 1 items - 30,70 €
Product Information
- Catalog #AvailabilityPrice
- Insert for screw cap GL 14, 6,0 mm inner diameterInsert for screw cap GL 14, 6,0 mm inner diameter554-3008Log in for availability1 * 1 items - 15,80 €