MultiScreen® PCR 384 Filter Plates
MultiScreen® PCR filter plates, Polystyrene, 100 μl, Proprietary membrane, 384, -
Catalog # MILFS384PCR50
Supplier: Merck
Jump to:
Product Details & Documents
- Colour code: Clear
- Purification of PCR products
- Research category
- Nucleic acid
- Ambient
Specifications
- Membrane:Proprietary membrane
- No. of wells:384
- Sterile:-
- Description:MultiScreen® PCR filter plates
- Material:Polystyrene
- Seq:02
- Well volume:100 μl



