Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, glass, with ground edges, 100 mm
Catalog # MURRG850/03
Supplier:   MURRAY S
Watch glasses
Catalog # MURRG850/03
Supplier:  MURRAY S
Watch glasses
Watch glass, glass, with ground edges, 100 mm
Price
53,10 €
Log in for availability
1 * 10 items
Product Details & Documents
Made of glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With ground edges for covering beakers.
Specifications
  • Ø:
    100 mm
  • Description:
    Watch glass, glass, with ground edges
  • Seq:
    03