Order Entry
Export
Orders LinkContactUsLinkComponent
Jars, tall

Jars, tall

60 ml, 45 mm, 76 mm
  MURRG805/03
 :   MURRAY S
Jars, tall
  MURRG805/03
 :  MURRAY S
Jars, tall
Jars, tall

60 ml, 45 mm, 76 mm

 
86,80 €
   
1 * 72 items
 

Clear or amber glass, without cap.

  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Amber coloured to protect light-sensitive materials

Perfect for sample collection, storing and transporting of powders, granulates, pastes or liquids.

 
  • Ext.Ø:
    45 mm
  • Capacity:
    60 ml
  • Height:
    76 mm
  • Seq:
    04