Order Entry
Export
Orders LinkContactUsLinkComponent
Bottles, sloping shoulder, round, with thread

Bottles, sloping shoulder, round, with thread

500 ml, R3/28, 89×54×202 mm
Catalog # MURRG802/08
Supplier:   MURRAY S
Bottles, sloping shoulder, round, with thread
Catalog # MURRG802/08
Supplier:  MURRAY S
Bottles, sloping shoulder, round, with thread

500 ml, R3/28, 89×54×202 mm

Price
221,00 €
Log in for availability
1 * 36 items
Product Details & Documents

Bottles, made of clear glass. Ideal for the sampling, storage and transportation of liquids.

  • Good chemical and physical properties
  • For use in applications without high thermal loads
  • Screw caps to be ordered separately
Specifications
  • Thread:
    R3/28
  • Capacity:
    500 ml
  • Seq:
    05
  • W×D×H:
    89×54×202 mm