Bottles, sloping shoulder, round, with thread
200 ml, R3/28, 63×37×175 mm
Catalog # MURRG802/06
Supplier: MURRAY S
Jump to:
Product Details & Documents
Bottles, made of clear glass. Ideal for the sampling, storage and transportation of liquids.
- Good chemical and physical properties
- For use in applications without high thermal loads
- Screw caps to be ordered separately
Specifications
- Thread:R3/28
- Capacity:200 ml
- Seq:03
- W×D×H:63×37×175 mm



