Order Entry
Export
Orders LinkContactUsLinkComponent
 

Dressing jars

200 mm, 250 mm
Catalog # MURRG500/09
Supplier:   MURRAY S
Dressing jars
Catalog # MURRG500/09
Supplier:  MURRAY S
Dressing jars

200 mm, 250 mm

Price
132,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Dressing jars, made of moulded glass, round.

  • Good chemical resistance
  • Lids must be ordered separately
Specifications
  • Ext.Ø:
    200 mm
  • Height:
    250 mm
  • Seq:
    09