Order Entry
Export
Orders LinkContactUsLinkComponent
Dressing jars

Dressing jars

100 mm, 150 mm
Catalog # MURRG500/02
Supplier:   MURRAY S
Dressing jars
Catalog # MURRG500/02
Supplier:  MURRAY S
Dressing jars

100 mm, 150 mm

Price
43,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Dressing jars, made of moulded glass, round.

  • Good chemical resistance
  • Lids must be ordered separately
Specifications
  • Ext.Ø:
    100 mm
  • Height:
    150 mm
  • Seq:
    03