0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Tubes
Test Tubes
Test tubes, without rim, light wall
Test tubes, without rim, light wall
125 mm, 16 mm, 0,8 mm
MURRG021/35
:
MURRAY S
Test tubes, without rim, light wall
MURRG021/35
:
MURRAY S
Test tubes, without rim, light wall
125 mm, 16 mm, 0,8 mm
66,40 €
1 * 100 items
-
+
Glass, clear.
Excellent chemical resistance
Good temperature resistance
Ext.Ø:
16 mm
Wall thickness:
0,8 mm
Length:
125 mm