Magnetic stirring bars, with pivot ring
70,78 mm, 9,22 mm
Catalog # KART799
Supplier: KARTELL
Jump to:
Product Details & Documents
PTFE coated suitable for use in containers with irregular bottom.
- Angled surfaces create high turbulence even at low speed
- Pivot ring reduces the contact surface and fosters optimal stir bar positioning
- Suitable for foodstuff
Specifications
- Ø:9,22 mm
- Length:70,78 mm
- Seq:08



