Order Entry
Export
Orders LinkContactUsLinkComponent
Watch dishes

Watch dishes

Watch glass, PTFE, 153,92 mm
Catalog # KART647
Supplier:   KARTELL
Watch dishes
Catalog # KART647
Supplier:  KARTELL
Watch dishes
Watch dishes

Watch glass, PTFE, 153,92 mm

Price
56,70 €
Log in for availability
1 * 1 items
Product Details & Documents

Made of PTFE, white.

  • Very good chemical, heat and mechanical resistance
  • Heat resistant up to +120 °C
  • For use with foodstuff

For blends or for covering vessels.

Specifications
  • Ø:
    153,92 mm
  • Description:
    Watch glass, PTFE
  • Seq:
    05