Order Entry
Export
Orders LinkContactUsLinkComponent
Watch dishes

Watch dishes

Watch glass, PTFE, 52,76 mm
  KART643
 :   KARTELL
Watch dishes
  KART643
 :  KARTELL
Watch dishes
Watch dishes

Watch glass, PTFE, 52,76 mm

 
14,60 €
   
1 * 1 items
 

Made of PTFE, white.

  • Very good chemical, heat and mechanical resistance
  • Heat resistant up to +120 °C
  • For use with foodstuff

For blends or for covering vessels.

 
  • Ø:
    52,76 mm
  • Description:
    Watch glass, PTFE
  • Seq:
    01