Order Entry
Export
Orders LinkContactUsLinkComponent
 

Pestles, serrated

270 mm, 30 ml mortars, 6,5 mm
  KART6330
 :   KARTELL
Pestles, serrated
  KART6330
 :  KARTELL
Pestles, serrated
Pestles, serrated

270 mm, 30 ml mortars, 6,5 mm

 
114,00 €
   
1 * 1 items
 

Pestles used in the combination with borosilicate mortar for sample crushing. Ideal for tough materials such as tumours.

  • Head made of PTFE
  • Shaft made of stainless steel
 
  • Ø:
    6,5 mm
  • For:
    30 ml mortars
  • Length:
    270 mm
  • Seq:
    05