Overhead stirrer tools, anchor stirrers
Anchor stirrer, DURAN®, 150 LF, 20000 ml, 16 mm
LENZ06321594
Anchor stirrers with GL thread connection.
- Made of precision calibrated glass tube
- With ground and polished bearing surface
- Suitable for flange vessels
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Description:Anchor stirrer, DURAN®, 150 LF
- Shaft Ø:16 mm
- For container:20000 ml



