Order Entry
Export
Orders LinkContactUsLinkComponent
Overhead stirrer tools, anchor stirrers

Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 150 LF, 2000 ml, 16 mm
Catalog # LENZ06321573
Supplier:   Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Catalog # LENZ06321573
Supplier:  Lenz Laborglas
Overhead stirrer tools, anchor stirrers
Overhead stirrer tools, anchor stirrers

Anchor stirrer, DURAN®, 150 LF, 2000 ml, 16 mm

Price
142,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Anchor stirrers with GL thread connection.

  • Made of precision calibrated glass tube
  • With ground and polished bearing surface
  • Suitable for flange vessels

Documents

Specifications
  • Description:
    Anchor stirrer, DURAN®, 150 LF
  • Shaft Ø:
    16 mm
  • For container:
    2000 ml