Flange clamps
ball and flat flanges KF/PF 15
Catalog # 201-0120
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Plastic, black.
- Easy to assemble and disassemble
- Tough construction, good temperature resistance
- Matching gaskets are available as an option
For fast, secure connection of ball or flat flanges. No gasket.
Specifications
- For:ball and flat flanges KF/PF 15
- Seq:01