Orders LinkContactUsLinkComponent
Flat flange lids, centre neck
laboratory flange DN 150, 1 Neck, NS 45/40 (centre), 140 mm
  LENZ06100406
Flat flange lids, centre neck
  LENZ06100406
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratory flange DN 150, 1 Neck, NS 45/40 (centre), 140 mm
320,00 €
1 * 1 items
DURAN®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
  • Type:
    1 Neck
  • With:
    laboratory flange DN 150
  • Neck:
    NS 45/40 (centre)
  • Total height:
    140 mm