Flat flange lids, centre neck
laboratory flange DN 150, 1 Neck, NS 29/32 (centre), 125 mm
LENZ06100404
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
- Type:1 Neck
- With:laboratory flange DN 150
- Neck:NS 29/32 (centre)
- Total height:125 mm