Orders LinkContactUsLinkComponent
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratory flange DN 60, 1 Neck, NS 29/32 (centre), 85 mm
Catalog # LENZ06100104
Supplier:   Lenz Laborglas
Flat flange lids, centre neck
Catalog # LENZ06100104
Supplier:  Lenz Laborglas
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratory flange DN 60, 1 Neck, NS 29/32 (centre), 85 mm
Price
124,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
Specifications
  • Type:
    1 Neck
  • With:
    laboratory flange DN 60
  • Neck:
    NS 29/32 (centre)
  • Total height:
    85 mm