Order Entry
Export
Orders LinkContactUsLinkComponent
Test tubes with standard ground joints, conical bottom

Test tubes with standard ground joints, conical bottom

120 mm, 17 mm, 14/23 NS
Catalog # LENZ05705216
Supplier:   Lenz Laborglas
Test tubes with standard ground joints, conical bottom
Catalog # LENZ05705216
Supplier:  Lenz Laborglas
Test tubes with standard ground joints, conical bottom
Test tubes with standard ground joints, conical bottom

120 mm, 17 mm, 14/23 NS

Price
12,50 €
Log in for availability
1 * 1 items
Product Details & Documents

Centrifuge tubes, made of DURAN® borosilicate glass 3.3.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Documents

Specifications
  • Ext.Ø:
    17 mm
  • Socket size:
    14/23 NS
  • Length:
    120 mm
  • Seq:
    06