Order Entry
Export
Orders LinkContactUsLinkComponent
 

Test tubes with NS stopper

100 mm, 16 mm, 14/23 NS
  LENZ05700207
Test tubes with NS stopper
  LENZ05700207
Test tubes with NS stopper
Test tubes with NS stopper

100 mm, 16 mm, 14/23 NS

 
7,00 €
   
1 * 1 items
 

DURAN®, borosilicate glass, clear. With standard ground joint and hexagonal glass stopper.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
 
  • Ext.Ø:
    16 mm
  • Socket size:
    14/23 NS
  • Length:
    100 mm
  • Seq:
    01