Orders LinkContactUsLinkComponent
Extractor heads for specific light solvents
Extractor heads for specific light solvents
500 ml
Catalog # LENZ05432158
Supplier:   Lenz Laborglas
Extractor heads for specific light solvents
Catalog # LENZ05432158
Supplier:  Lenz Laborglas
Extractor heads for specific light solvents
Extractor heads for specific light solvents
500 ml
Price
329,00 €
Log in for availability
1 * 1 items
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).
Documents
Specifications
  • Extraction cap.:
    500 ml
  • Seq:
    03