Order Entry
Export
Orders LinkContactUsLinkComponent
 

Extractors for specific light solvents

Complete set, 500 ml, 1000 ml, 60/46 NS
Catalog # LENZ05432058
Supplier:   Lenz Laborglas
Extractors for specific light solvents
Catalog # LENZ05432058
Supplier:  Lenz Laborglas
Extractors for specific light solvents
Extractors for specific light solvents

Complete set, 500 ml, 1000 ml, 60/46 NS

Price
647,00 €
Log in for availability
1 * 1 items
Product Details & Documents

DURAN®, Borosilicate glass.

  • Very good chemical resistance
  • Very good temperature resistance

For the extraction of liquids with specific light solvents.

Specifications
  • Extraction cap.:
    500 ml
  • Socket size:
    60/46 NS
  • Type:
    Complete set
  • Flask capacity:
    1000 ml