Twisselmann extractors
Head only, 30 ml, 29/32 NS
Catalog # LENZ05415125
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Extraction cap.:30 ml
- Socket size:29/32 NS
- Type:Head only