Twisselmann extractors
Complete, 30 ml, 100 ml, 29/32 NS
LENZ05380025
DURAN®, borosilicate glass.
- Very good chemical resistance
- Very good temperature resistance
- Extraction cap.:30 ml
- Socket size:29/32 NS
- Type:Complete
- Flask capacity:100 ml
DURAN®, borosilicate glass.