Orders LinkContactUsLinkComponent
Cold traps with/without condensation drain
Cold traps with/without condensation drain
1000 ml, with condensate drain
Catalog # LENZ05355570
Supplier:   Lenz Laborglas
Cold traps with/without condensation drain
Catalog # LENZ05355570
Supplier:  Lenz Laborglas
Cold traps with/without condensation drain
Cold traps with/without condensation drain
1000 ml, with condensate drain
Price
1 120,00 €
Log in for availability
1 * 1 items
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump. DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
Specifications
  • Description:
    1000 ml, with condensate drain
  • Seq:
    06