Cold traps with/without condensation drain
1000 ml, without condensate drain
Catalog # LENZ05355070
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
For direct filling with coolant. Double-walled design ensures low coolant consumption. Includes NS 29/32 standard ground socket for connecting to the apparatus and NS 29/32 standard ground cone for connecting to the vacuum pump. DURAN®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Description:1000 ml, without condensate drain
- Seq:03