Order Entry
Export
Orders LinkContactUsLinkComponent
 

Funnels with short stem

60 mm, 8 mm, 60 mm
  LENZ05318017
Funnels with short stem
  LENZ05318017
Funnels with short stem
Funnels with short stem

60 mm, 8 mm, 60 mm

 
4,40 €
   
1 * 1 items
 

Soda-lime glass.

  • Excellent chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean

With smooth internal walls. Flare angle 60°.

 
  • Funnel O-Ø:
    60 mm
  • Stem O-Ø:
    8 mm
  • Stem length:
    60 mm