Order Entry
Export
Orders LinkContactUsLinkComponent
 

Funnels with short stem

35 mm, 6 mm, 35 mm
  LENZ05318005
Funnels with short stem
  LENZ05318005
Funnels with short stem
Funnels with short stem

35 mm, 6 mm, 35 mm

 
30,00 €
   
1 * 10 items
 

Soda-lime glass.

  • Excellent chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean

With smooth internal walls. Flare angle 60°.

 
  • Funnel O-Ø:
    35 mm
  • Stem O-Ø:
    6 mm
  • Stem length:
    35 mm